Learn More
RP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RP1.
Brand: Abnova PAB23915.100uL
Description
Initially named for its response to in vivo retinal oxygen levels (designated ORP1 for 'oxygen-regulated protein-1'), this gene was subsequently linked to autosomal dominant retinitis pigmentosa and was renamed RP1 for 'retinitis pigmentosa 1'. The data suggest that mutations in this gene cause dominant RP, and that the encoded protein has an important but unknown role in photoreceptor biology. There is evidence for at least one transcript variant that overlaps the current reference sequence and which has several exons upstream and downstream of the current reference sequence. However, the full-length nature of this variant cannot be determined at this time. [provided by RefSeq]
Sequence: VTCSPCEMCTVNKAYSPKETCNPSDTFFPSDGYGVDQTSMNKACFLGEVCSLTDTVFSDKACAQKENHTYEGACPIDETYVPVNVCNTIDFLNSKENTYTDNLDSTEELERGDDIQKDLNILTDPEYKNGFNTLVSHQNVSNLSSCGSpecifications
| RP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RP1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DCDC4A/ORP1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6101 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
| RP1 | |
| RP1 | |
| Recombinant protein corresponding to amino acids of human RP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only