missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPN3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RPN3.
Brand: Abnova PAB15593.100uL
This item is not returnable.
View return policy
Specifications
| RPN3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RPN3. | |
| In antiserum (0.05% sodium azide) | |
| SUN2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody react with S. cerevisiae RPN3. | |
| Store at 4°C. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| RPN3 | |
| RPN3 | |
| Recombinant protein corresponding to Saccharomyces cerevisiae RPN3. | |
| RUO | |
| 856742 | |
| Yeast | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction