Learn More
RPS6KA4 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RPS6KA4.
Brand: Abnova PAB4524.400uL
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq
Specifications
| RPS6KA4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of RPS6KA4. | |
| In PBS (0.09% sodium azide) | |
| MSK2/RSK-B | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 8986 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunofluorescence (1:100-500) The optimal working dilution should be determined by the end user. | |
| RPS6KA4 | |
| RPS6KA4 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human RPS6KA4. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only