Learn More
RSPO3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RSPO3.
Brand: Abnova PAB22374.100uL
Description
This gene encodes a member of the thrombospondin type 1 repeat supergene family. In addition, the protein contains a furin-like cysteine-rich region. Furin-like repeat domains have been found in a variety of eukaryotic proteins involved in the mechanism of signal transduction by receptor tyrosine kinases. [provided by RefSeq
Sequence: PTNETRKCTVQRKKCQKGERGKKGRERKRKKPNKGESKEAIPDSKSLESSKEIPEQRENKQQQKKRKVQDKQKSVSVSSpecifications
| RSPO3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RSPO3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CRISTIN1/FLJ14440/PWTSR/THSD2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 84870 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| RSPO3 | |
| RSPO3 | |
| Recombinant protein corresponding to amino acids of human RSPO3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only