missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXFP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RXFP1.
Brand: Abnova PAB16536.100uL
This item is not returnable.
View return policy
Description
Sequence: RFWYNYRQRKSMDSKGQKSpecifications
| RXFP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of RXFP1. | |
| In serum (50% glycerol, 0.025% sodium azide) | |
| LGR7/LGR7.1/LGR7.10/LGR7.2/MGC138347/MGC142177/RXFPR1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform 2 and isoform 4). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user. | |
| RXFP1 | |
| RXFP1 | |
| A synthetic peptide corresponding to amino acids 694-711 of human RXFP1. | |
| RUO | |
| 59350 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction