Learn More
S1PR3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of S1PR3.
Brand: Abnova PAB26238.50ug
Description
This gene encodes a member of the EDG family of receptors, which are G protein-coupled receptors. This protein has been identified as a functional receptor for sphingosine 1-phosphate and likely contributes to the regulation of angiogenesis and vascular endothelial cell function. [provided by RefSeq
Specifications
| S1PR3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of S1PR3. | |
| In PBS (0.09% sodium azide) | |
| EDG-3/EDG3/FLJ37523/FLJ93220/LPB3/MGC71696/S1P3 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 1903 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| S1PR3 | |
| S1PR3 | |
| A synthetic peptide corresponding to 19 amino acid at N-terminus of human S1PR3. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except NAA16 (53%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only