missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sad Rabbit anti-Fruit fly, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of sad.
Brand: Abnova PAB8990.100ug
This item is not returnable.
View return policy
Description
Sequence: GRYLPQDAQLGGHFIEKDSpecifications
| sad | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of sad. | |
| In buffer containing 0.02% sodium azide | |
| 315a1/CG14728/CYP315A1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Fruitfly | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| sad | |
| sad | |
| A synthetic peptide corresponding to fruit fly sad. | |
| RUO | |
| 44858 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction