missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARB1 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Scarb1.
Brand: Abnova PAB12042.100uL
This item is not returnable.
View return policy
Description
Sequence: CSPAAKGTVLQEAKL-COOHSpecifications
| SCARB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Scarb1. | |
| In antiserum | |
| AI120173/CD36/Cd36l1/Cla-1/Cla1/D5Ertd460e/SR-B1/SR-BI/SRBI/Srb1/mSR-BI | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is specific to SR-BI. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Immunocytochemistry (1:50-1:1000) Immunofluorescence (1:50-1:1000) Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user. | |
| Scarb1 | |
| Scarb1 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 496-509 of mouse Scarb1. | |
| RUO | |
| 20778 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction