Learn More
SELK, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SELK.
Brand: Abnova PAB25648.100ug
Description
This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This selenoprotein is localized to the endoplasmic reticulum and is highly expressed in the heart, where it may function as an antioxidant. [provided by RefSeq]
Specifications
| SELK | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| SELK | |
| SELK | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human SELK. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of SELK. | |
| In PBS (0.05% sodium azide) | |
| HSPC030/HSPC297/MGC17057 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 58515 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only