Learn More
SEMA3B Rabbit anti-Bovine, Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SEMA3B.
Brand: Abnova PAB12040.50uL
Description
The semaphorin/collapsin family of molecules plays a critical role in the guidance of growth cones during neuronal development. The secreted protein encoded by this gene family member is important in axonal guidance and has been shown to act as a tumor suppressor by inducing apoptosis. [provided by RefSeq
Specifications
| SEMA3B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SEMA3B. | |
| In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
| FLJ34863/LUCA-1/SEMA5/SEMAA/SemA/semaV | |
| Rabbit | |
| 50 μL | |
| Primary | |
| This antibody is specific to SEMA 3B. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:5000-1:20000) The optimal working dilution should be determined by the end user. | |
| SEMA3B | |
| SEMA3B | |
| A synthetic peptide corresponding to amino acids 100-200 of human SEMA3B. | |
| RUO | |
| 7869 | |
| Bovine, Canine, Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only