Learn More
SETDB2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SETDB2.
Brand: Abnova PAB5573.100ug
Description
Proteins that contain a SET domain, such as SETDB2, modulate gene expression epigenetically through histone H3 (see MIM 601128) methylation. SETDB2 is likely a histone H3 methyltransferase, as it contains both the active site and flanking cysteine residues required for catalytic activity (Zhang et al., 2003 [PubMed 12754510]).[supplied by OMIM
Specifications
| SETDB2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SETDB2. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| C13orf4/CLLD8/CLLL8/DKFZp586I0123/DKFZp761J1217/KMT1F | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| SETDB2 | |
| SETDB2 | |
| A synthetic peptide corresponding to N-terminus of human SETDB2. | |
| RUO | |
| 83852 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only