missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SF1.
Brand: Abnova PAB18220.100ug
This item is not returnable.
View return policy
Specifications
| SF1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| SF1 | |
| SF1 | |
| A synthetic peptide corresponding to residues surrounding S82 of human SF1. | |
| 100 μg | |
| Primary | |
| This antibody is specific to SF1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of SF1. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| D11S636/ZFM1/ZNF162 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 7536 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction