Learn More
SGK3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SGK3.
Brand: Abnova PAB2320.400uL
Description
This gene is a member of the Ser/Thr protein kinase family and encodes a phosphoprotein with a PX (phox homology) domain. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
| SGK3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SGK3. | |
| In PBS (0.09% sodium azide) | |
| CISK/DKFZp781N0293/SGK2/SGKL | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 23678 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| SGK3 | |
| SGK3 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human SGK3. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only