missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Sheep IgM Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB7206-1MG
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal specifically detects IgM Heavy Chain in Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IgM | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Sheep IgM | |
| 1 mg | |
| Secondary | |
| By immunoelectrophoresis and ELISA this reacts specifically with sheep IgM. This may cross react with IgM from other species. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1:1000 - 1:10000, ELISA, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3507 | |
| Sheep | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction