Learn More
SIGLEC11 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC11.
Brand:  Abnova PAB3646.400uL
Product Code. 16038757
  • € 388.00 / 400µL
Description
SIGLECs are a family of cell surface lectins defined by shared structural motifs in the first 2 immunoglobulin (Ig)-like domains and by their ability to recognize sialic acids via the first Ig V set domain.[supplied by OMIM
Specifications
Specifications
SIGLEC11
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC11.
In PBS (0.09% sodium azide)
SIGLEC11
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human SIGLEC11.
400 μL
Primary
Human
Liquid
ELISA, Immunohistochemistry, Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
SIGLEC11
Rabbit
Protein A purification
RUO
114132
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.