Learn More
SIRT5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SIRT5.
Brand: Abnova PAB4825.400uL
Descrizione
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class III of the sirtuin family. Alternative splicing of this gene results in two transcript variants. [provided by RefSeq
Specifica
| SIRT5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SIRT5. | |
| In PBS (0.09% sodium azide) | |
| FLJ36950/SIR2L5 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 23408 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| SIRT5 | |
| SIRT5 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human SIRT5. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only