Learn More
SIRT7 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SIRT7.
Brand: Abnova PAB0197.100ug
Description
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class IV of the sirtuin family. [provided by RefSeq
Specifications
| SIRT7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SIRT7. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| MGC126840/MGC126842/SIR2L7 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| SIRT7 | |
| SIRT7 | |
| A mixture of synthetic peptides corresponding to amino acids 35-51 and 361-377 of human SIRT7. | |
| RUO | |
| 51547 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only