Learn More
SKA3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SKA3.
Brand: Abnova PAB16834.100ug
Description
This gene encodes a component of the spindle and kinetochore-associated protein complex that regulates microtubule attachment to the kinetochores during mitosis. The encoded protein localizes to the outer kinetechore and may be required for normal chromosome segregation and cell division. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Specifications
| SKA3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SKA3. | |
| In PBS (0.02% sodium azide) | |
| NP_659498 | |
| C13orf3 | |
| A synthetic peptide corresponding to C-terminus 13 amino acids of human SKA3. | |
| RUO | |
| 221150 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| C13orf3 | |
| MGC4832/RAMA1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only