Learn More
SLAIN2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLAIN2.
Brand:  Abnova PAB25660.100ug
Product Code. 16005837
  • € 619.00 / 100µg
Specifications
SLAIN2
Polyclonal
Unconjugated
Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
SLAIN2
SLAIN2
A synthetic peptide corresponding to 14 amino acids at N-terminus of human SLAIN2.
100 μg
Primary
SLAIN2 antibody is predicted not to cross-react with SLAIN1.
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
ELISA, Immunohistochemistry
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of SLAIN2.
In PBS (0.05% sodium azide)
FLJ21611/KIAA1458
Rabbit
Peptide affinity purification
RUO
57606