missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slc14a2, Rabbit, ATTO488, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.
Brand: Abnova PAB28822.100ug
This item is not returnable.
View return policy
Specifications
| Slc14a2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant Rat Slc14a2. | |
| PBS, 50% glycerol and 0.09% sodium azide | |
| Slc14a2T/UrT1-C/UrT1-D/testSymbol | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 54302 | |
| Mouse, Rat | |
| Liquid |
| Immunofluorescence, Western Blot | |
| Atto 488 | |
| Immunofluorescence Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Slc14a2 | |
| Slc14a2 | |
| Recombinant protein corresponding to Slc14a2 C-terminus (amino acids 911-929). | |
| 100 μg | |
| Primary | |
| 1 ug/mL of PAB28822 was sufficient for detection of Slc14a2, ∼97 and 127kDa, in 20ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction