missing translation for 'onlineSavingsMsg'
Learn More

Slc14a2, Rabbit, PerCP, Polyclonal Antibody, Abnova™

Product Code. 16253980
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16253980 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16253980 Supplier Abnova Supplier No. PAB28835.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.

Specifications

Antigen Slc14a2
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate PerCP
Description Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.
Dilution Immunofluorescence Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation PBS, 50% glycerol and 0.09% sodium azide
Gene Slc14a2
Gene Alias Slc14a2T/UrT1-C/UrT1-D/testSymbol
Gene Symbols Slc14a2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Slc14a2 C-terminus (amino acids 911-929).
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54302
Test Specificity 1 ug/mL of PAB28835 was sufficient for detection of Slc14a2, ∼97 and 127kDa, in 20ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody.
Target Species Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.