Learn More
SLC16A3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLC16A3.
Brand: Abnova PAB17775.100ug
Description
Lactic acid and pyruvate transport across plasma membranes is catalyzed by members of the proton-linked monocarboxylate transporter (MCT) family, which has been designated solute carrier family-16. Each MCT appears to have slightly different substrate and inhibitor specificities and transport kinetics, which are related to the metabolic requirements of the tissues in which it is found. The MCTs, which include MCT1 (SLC16A1; MIM 600682) and MCT2 (SLC16A7; MIM 603654), are characterized by 12 predicted transmembrane domains (Price et al., 1998 [PubMed 9425115]).[supplied by OMIM
Specifications
| SLC16A3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SLC16A3. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| MCT3/MCT4/MGC138472/MGC138474 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total SLC16A3 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| SLC16A3 | |
| SLC16A3 | |
| A synthetic peptide corresponding to internal of human SLC16A3. | |
| RUO | |
| 9123 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only