Learn More
SLC22A11, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLC22A11.
Brand: Abnova PAB8458.20ug
Description
The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and is found mainly in the kidney and in the placenta, where it may act to prevent potentially harmful organic anions from reaching the fetus. [provided by RefSeq
Specifications
| SLC22A11 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SLC22A11. | |
| In PBS (0.1% proclin, 2.0% Block Ace) | |
| MGC34282/OAT4/hOAT4 | |
| Rabbit | |
| 20 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Immunohistochemistry (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SLC22A11 | |
| SLC22A11 | |
| A synthetic peptide corresponding to human SLC22A11. | |
| RUO | |
| 55867 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only