missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC43A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SLC43A1.
Brand: Abnova PAB21048.100uL
This item is not returnable.
View return policy
Description
SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids (Babu et al., 2003 [PubMed 12930836]).[supplied by OMIM
Sequence: NCTLNWPIEAFPAPEEVNYTKKIKLSGLALDHKVTGDLFYTHVTTMGQRLSQKAPSLEDGSDAFMSPQDVRGTSENLPERSVPLRKSLCSSpecifications
| SLC43A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SLC43A1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| LAT3/PB39/POV1/R00504 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8501 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| SLC43A1 | |
| SLC43A1 | |
| Recombinant protein corresponding to amino acids of human SLC43A1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto