missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A1 Rabbit anti-Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Slc6a1.
Brand: Abnova PAB14540.100uL
This item is not returnable.
View return policy
Specifications
| SLC6A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Slc6a1. | |
| In serum | |
| Gabt1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 79212 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:100-1:200) The optimal working dilution should be determined by the end user. | |
| Slc6a1 | |
| Slc6a1 | |
| A synthetic peptide corresponding to C-terminus rat Slc6a1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction