missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A15, Rabbit, Polyclonal Antibody, Abnova™
Description
SLC6A15 shows structural characteristics of an Na(+) and Cl(-)-dependent neurotransmitter transporter, including 12 transmembrane (TM) domains, intracellular N and C termini, and large extracellular loops containing multiple N-glycosylation sites (Farmer et al., 2000 [PubMed 11112352]).[supplied by OMIM
Sequence: RELDDDVTESVKDLLSNEDAADDAFKTSELIVDGQEEKDTDVEEGSEVEDERPAWNSKLQ
Specifications
Specifications
| Antigen | SLC6A15 |
| Applications | Immunohistochemistry (PFA fixed) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against recombinant SLC6A15. |
| Dilution | Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) |
| Gene | SLC6A15 |
| Gene Alias | DKFZp761I0921/FLJ10316/MGC87066/NTT73/SBAT1/V7-3/hv7-3 |
| Gene Symbols | SLC6A15 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?