missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMT3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SMT3.
Brand: Abnova PAB11357.500ug
This item is not returnable.
View return policy
Specifications
| SMT3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SMT3. | |
| Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| SMT3 | |
| Recombinant protein corresponding to Saccharomyces cerevisiae SMT3. | |
| RUO | |
| 852122 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000-1:5000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| SMT3 | |
| Rabbit | |
| 500 μg | |
| Primary | |
| Yeast | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction