Learn More
SNAI1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SNAI1.
Brand: Abnova PAB1924.400uL
Description
The Drosophila embryonic protein snail is a zinc finger transcriptional repressor which downregulates the expression of ectodermal genes within the mesoderm. The nuclear protein encoded by this gene is structurally similar to the Drosophila snail protein, and is also thought to be critical for mesoderm formation in the developing embryo. At least two variants of a similar processed pseudogene have been found on chromosome 2. [provided by RefSeq
Specifications
| SNAI1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SNAI1. | |
| In PBS (0.09% sodium azide) | |
| SLUGH2/SNA/SNAH/dJ710H13.1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 6615 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| SNAI1 | |
| SNAI1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human SNAI1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only