Learn More
SNAI2 (phospho S155/S158/S160/Y164/Y169), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Description
This gene encodes a member of the Snail family of C2H2-type zinc finger transcription factors. The encoded protein acts as a transcriptional repressor that binds to E-box motifs and is also likely to repress E-cadherin transcription in breast carcinoma. This protein is involved in epithelial-mesenchymal transitions and has antiapoptotic activity. Mutations in this gene may be associated with sporatic cases of neural tube defects. [provided by RefSeq
Specifications
Specifications
| Antigen | SNAI2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic phosphopeptide of SNAI2. |
| Dilution | ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) |
| Gene | SNAI2 |
| Gene Alias | MGC10182/SLUG/SLUGH1/WS2D |
| Gene Symbols | SNAI2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.