Learn More
SOCS1 Rabbit anti-Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SOCS1.
Brand: Abnova PAB8011.100ug
Description
This gene encodes a member of the STAT-induced STAT inhibitor (SSI), also known as suppressor of cytokine signaling (SOCS), family. SSI family members are cytokine-inducible negative regulators of cytokine signaling. The expression of this gene can be induced by a subset of cytokines, including IL2, IL3 erythropoietin (EPO), CSF2/GM-CSF, and interferon (IFN)-gamma. The protein encoded by this gene functions downstream of cytokine receptors, and takes part in a negative feedback loop to attenuate cytokine signaling. Knockout studies in mice suggested the role of this gene as a modulator of IFN-gamma action, which is required for normal postnatal growth and survival. [provided by RefSeq
Specifications
| SOCS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SOCS1. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| CIS1/CISH1/JAB/SOCS-1/SSI-1/SSI1/TIP3 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 8651 | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| SOCS1 | |
| SOCS1 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human SOCS1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rabbit, Rat | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only