Learn More
SOD1 Rabbit anti-Bovine, Dog, Frog, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against SOD1.
Brand: Abnova PAB14492.100uL
Description
The protein encoded by this gene binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. The encoded isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene. [provided by RefSeq
Specifications
| SOD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against SOD1. | |
| In PBS, pH 7.0 (50% glycerol, 0.09% sodium azide) | |
| ALS/ALS1/IPOA/SOD/homodimer | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody detects a 23 KDa protein in human (and a 19 KDa protein in other species) on SDS-PAGE immunoblots, corresponding to the molecular mass of Cu/Zn SOD. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.2 ug/mL) Immunohistochemistry (1 ug/mL) Immunoprecipitation (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| SOD1 | |
| SOD1 | |
| Human SOD1 (Cu/Zn Superoxide dismutase). | |
| RUO | |
| 6647 | |
| Bovine, Canine, Frog, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only