Learn More
SOX9 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SOX9.
Brand: Abnova PAB12737.100ug
Description
The protein encoded by this gene recognizes the sequence CCTTGAG along with other members of the HMG-box class DNA-binding proteins. It acts during chondrocyte differentiation and, with steroidogenic factor 1, regulates transcription of the anti-Muellerian hormone (AMH) gene. Deficiencies lead to the skeletal malformation syndrome campomelic dysplasia, frequently with sex reversal. [provided by RefSeq
Specifications
| SOX9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SOX9. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| CMD1/CMPD1/SRA1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼56 KDa of human SOX9 without phosphorylation at S181. It does not recognize the phosphorylation protein at Swe 181. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SOX9 | |
| SOX9 | |
| A synthetic peptide corresponding to residues surrounding S181 (RKSVK) of human SOX9. | |
| RUO | |
| 6662 | |
| Human, Mouse, Rat | |
| Liquid |