Learn More
SOX9 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SOX9.
Brand:  Abnova PAB12737.100ug
Product Code. 16090107
  • € 388.00 / 100µg
Description
The protein encoded by this gene recognizes the sequence CCTTGAG along with other members of the HMG-box class DNA-binding proteins. It acts during chondrocyte differentiation and, with steroidogenic factor 1, regulates transcription of the anti-Muellerian hormone (AMH) gene. Deficiencies lead to the skeletal malformation syndrome campomelic dysplasia, frequently with sex reversal. [provided by RefSeq
Specifications
Specifications
SOX9
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of SOX9.
In TBS, pH 7.2 (BSA, 10% Proclin300)
CMD1/CMPD1/SRA1
Rabbit
100 μg
Primary
This antibody recognizes ∼56 KDa of human SOX9 without phosphorylation at S181. It does not recognize the phosphorylation protein at Swe 181.
Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user.
SOX9
SOX9
A synthetic peptide corresponding to residues surrounding S181 (RKSVK) of human SOX9.
RUO
6662
Human, Mouse, Rat