Learn More
SPTBN1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SPTBN1.
Brand: Abnova PAB27161.100uL
Description
Spectrin is an actin crosslinking and molecular scaffold protein that links the plasma membrane to the actin cytoskeleton, and functions in the determination of cell shape, arrangement of transmembrane proteins, and organization of organelles. It is composed of two antiparallel dimers of alpha- and beta- subunits. This gene is one member of a family of beta-spectrin genes. The encoded protein contains an N-terminal actin-binding domain, and 17 spectrin repeats which are involved in dimer formation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| SPTBN1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| SPTBN1 | |
| SPTBN1 | |
| A synthetic peptide corresponding to human SPTBN1. | |
| 100 μL | |
| Primary | |
| SPTBN1 polyclonal antibody detects endogenous levels of SPTBN1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of SPTBN1. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| ELF/SPTB2/betaSpII | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6711 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only