Learn More
SQSTM1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SQSTM1.
Brand: Abnova PAB1750.400uL
Description
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B (NF-kB) signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NFkb in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone. [provided by RefSeq
Specifications
| SQSTM1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SQSTM1. | |
| In PBS (0.09% sodium azide) | |
| A170/OSIL/PDB3/ZIP3/p60/p62/p62B | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 8878 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) Immunofluorescence (1:50-100) The optimal working dilution should be determined by the end user. | |
| SQSTM1 | |
| SQSTM1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human SQSTM1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only