Learn More
ST13 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ST13.
Brand: Abnova PAB4827.400uL
Description
The protein encoded by this gene is an adaptor protein that mediates the association of the heat shock proteins HSP70 and HSP90. This protein has been shown to be involved in the assembly process of glucocorticoid receptor, which requires the assistance of multiple molecular chaperones. The expression of this gene is reported to be downregulated in colorectal carcinoma tissue suggesting that is a candidate tumor suppressor gene. [provided by RefSeq
Specifications
| ST13 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ST13. | |
| In PBS (0.09% sodium azide) | |
| AAG2/FAM10A1/FAM10A4/FLJ27260/HIP/HOP/HSPABP/HSPABP1/MGC129952/P48/PRO0786/SNC6 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 6767 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
| ST13 | |
| ST13 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ST13. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only