Learn More
STAT1 (phospho S727), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of STAT1.
Brand: Abnova PAB0613.400uL
Description
The protein encoded by this gene is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein can be activated by various ligands including interferon-alpha, interferon-gamma, EGF, PDGF and IL6. This protein mediates the expression of a variety of genes, which is thought to be important for cell viability in response to different cell stimuli and pathogens. Two alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq
Specifications
| STAT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of STAT1. | |
| In PBS (0.09% sodium azide) | |
| DKFZp686B04100/ISGF-3/STAT91 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 6772 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| STAT1 | |
| STAT1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S727 of human STAT1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only