missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5A Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a synthetic peptide of Stat5a.
Brand: Abnova PAB0834.200ug
This item is not returnable.
View return policy
Specifications
| STAT5A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a synthetic peptide of Stat5a. | |
| In PBS (0.2% gelatin, 0.09% sodium azide) | |
| AA959963/STAT5 | |
| Rabbit | |
| 200 μg | |
| Primary | |
| This antibody does not cross-react with Stat5b. | |
| Store at 4°C. Do not freeze. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:200) The optimal working dilution should be determined by the end user. | |
| Stat5a | |
| Stat5a | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of mouse Stat5a. | |
| RUO | |
| 20850 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction