missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRN Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STRN.
Brand: Abnova PAB12016.100uL
This item is not returnable.
View return policy
Specifications
| STRN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of STRN. | |
| In antiserum (0.02% sodium azide) | |
| MGC125642/SG2NA | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody does not cross-react with SG2NA. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Immunofluorescence (1:5000) Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user. | |
| STRN | |
| STRN | |
| A synthetic peptide corresponding to amino acids 350-400 of human STRN. | |
| RUO | |
| 6801 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction