missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYCP3 Rabbit anti-Human, Mouse, Biotin, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SYCP3.
Brand: Abnova PAB15243.100uL
This item is not returnable.
View return policy
Specifications
| SYCP3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SYCP3. | |
| In PBS | |
| COR1/MGC71888/SCP3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This SCP3 antibody recognizes human and murine protein. Other species have not been tested. | |
| Store at 4°C. Do not freeze. |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Biotin | |
| Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SYCP3 | |
| SYCP3 | |
| A synthetic peptide corresponding to C-terminus of human SYCP3. | |
| RUO | |
| 50511 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction