Learn More
SYNGR2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SYNGR2.
Brand: Abnova PAB28183.50ug
Beschreibung
This gene encodes an integral membrane protein containing four transmembrane regions and a C-terminal cytoplasmic tail that is tyrosine phosphorylated. The exact function of this protein is unclear, but studies of a similar rat protein suggest that it may play a role in regulating membrane traffic in non-neuronal cells. The gene belongs to the synaptogyrin gene family. [provided by RefSeq
Spezifikation
| SYNGR2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SYNGR2. | |
| In PBS (0.09% sodium azide) | |
| MGC102914 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 9144 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/ml) | |
| SYNGR2 | |
| SYNGR2 | |
| A synthetic peptide corresponding to 17 amino acids at N-terminus region of human SYNGR2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only