Learn More
TACR1, Rabbit, DyLight 549, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TACR1.
Brand: Abnova PAB15187.100uL
Description
This gene belongs to a gene family of tachykinin receptors. These tachykinin receptors are characterized by interactions with G proteins and contain seven hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin substance P, also referred to as neurokinin 1. The encoded protein is also involved in the mediation of phosphatidylinositol metabolism of substance P. [provided by RefSeq
Specifications
| TACR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TACR1. | |
| In 50mM sodium borate | |
| NK1R/NKIR/SPR/TAC1R | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This is specific to human NK-1. This product may be used for Western blotting where a band is seen at ∼53 KDa representing human NK-1. The actual molecular weight may vary depending upon the tissue used. | |
| Store at 4°C. Do not freeze. |
| Flow Cytometry, Immunohistochemistry | |
| DyLight 549 | |
| Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| TACR1 | |
| TACR1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human TACR1. | |
| RUO | |
| 6869 | |
| Human, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only