missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACR3 Rabbit anti-Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Tacr3.
Brand: Abnova PAB0710.100uL
This item is not returnable.
View return policy
Description
Sequence: ASTTSSFISSPYTSVDEYSSpecifications
| TACR3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Tacr3. | |
| Lyophilized from PBS | |
| Nkr/Nmkr/Tac3r | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Rat | |
| Lyophilized |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:2000) The optimal dilution should be determined by the end user. | |
| Tacr3 | |
| Tacr3 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 434-452 of rat Tacr3. | |
| RUO | |
| 24808 | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction