Learn More
TADA2A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TADA2A.
Brand: Abnova PAB17345.100ug
Description
Many DNA-binding transcriptional activator proteins enhance the initiation rate of RNA polymerase II-mediated gene transcription by interacting functionally with the general transcription machinery bound at the basal promoter. Adaptor proteins are usually required for this activation, possibly to acetylate and destabilize nucleosomes, thereby relieving chromatin constraints at the promoter. The protein encoded by this gene is a transcriptional activator adaptor and has been found to be part of the PCAF histone acetylase complex. Two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq
Specifications
| TADA2A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TADA2A. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| ADA2/ADA2A/FLJ12705/KL04P/hADA2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total TADA2A protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| TADA2L | |
| TADA2L | |
| A synthetic peptide corresponding to internal of human TADA2A. | |
| RUO | |
| 6871 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only