Learn More
TBC1D1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TBC1D1.
Brand: Abnova PAB12998.100ug
Description
TBC1D1 is the founding member of a family of proteins sharing a 180- to 200-amino acid TBC domain presumed to have a role in regulating cell growth and differentiation. These proteins share significant homology with TRE2 (USP6; MIM 604334), yeast Bub2, and CDC16 (MIM 603461) (White et al., 2000 [PubMed 10965142]).[supplied by OMIM
Specifications
| TBC1D1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TBC1D1. | |
| In PBS (0.02% sodium azide) | |
| KIAA1108/TBC/TBC1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| TBC1D1 | |
| TBC1D1 | |
| A synthetic peptide corresponding to C-terminus 22 amino acids of human TBC1D1. | |
| RUO | |
| 23216 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only