Learn More
TBKBP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TBKBP1.
Brand: Abnova PAB16831.100ug
Description
TBKBP1 is an adaptor protein that binds to TBK1 (MIM 604834) and is part of the interaction network in the TNF (MIM 191160)/NFKB (see MIM 164011) pathway (Bouwmeester et al., 2004 [PubMed 14743216]).[supplied by OMIM
Specifications
| TBKBP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TBKBP1. | |
| In PBS (0.02% sodium azide) | |
| KIAA0775/ProSAPiP2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| TBKBP1 | |
| TBKBP1 | |
| A synthetic peptide corresponding to C-terminus 16 amino acids of human TBKBP1. | |
| RUO | |
| 9755 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only