Learn More
TCEB3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TCEB3.
Brand: Abnova PAB3716.400uL
Description
This gene encodes the protein elongin A, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. [provided by RefSeq
Specifications
| TCEB3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TCEB3. | |
| In PBS (0.09% sodium azide) | |
| FLJ38760/FLJ42849/SIII/TCEB3A | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 6924 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| TCEB3 | |
| TCEB3 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human TCEB3. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only