Learn More
TFAP4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TFAP4.
Brand: Abnova PAB28535.100uL
Description
Transcription factors of the basic helix-loop-helix-zipper (bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization. Transcription factor AP4 activates both viral and cellular genes by binding to the symmetrical DNA sequence CAGCTG (Mermod et al., 1988 [PubMed 2833704]; Hu et al., 1990 [PubMed 2123466]).[supplied by OMIM
Sequence: YFMVPTQKVPSLQHFRKTEKEVIGGLCSLANIPLTPETQRDQERRIRREIANSNERRRMQSINAGFQSLKTLIPHTDGEKLSKAAILQQTAEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEKSpecifications
| TFAP4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TFAP4. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| AP-4/bHLHc41 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7023 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:250) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| TFAP4 | |
| TFAP4 | |
| Recombinant protein corresponding to amino acids of human TFAP4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only