missing translation for 'onlineSavingsMsg'
Learn More

TGFB2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Product Code. 16010117
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16010117 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16010117 Supplier Abnova Supplier No. PAB12739.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of TGFB2.

Specifications

Antigen TGFB2
Applications ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of TGFB2.
Dilution Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Gene TGFB2
Gene Alias MGC116892/TGF-beta2
Gene Symbols TGFB2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human TGFB2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7042
Test Specificity This antibody recognizes ∼13 KDa of TGFB2, not TGFB1 and less extense of TGFB3.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. Upon initial thawing apportion into working aliquots and store at -80°C for up to six months.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.