Learn More
TICAM1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TICAM1.
Brand: Abnova PAB12905.100ug
Description
TRIF, or TICAM1, is a Toll/IL1R (MIM 147810) (TIR) domain-containing adaptor molecule, like MYD88 (MIM 602170) and TIRAP (MIM 606252), that induces interferon-beta (IFNB; MIM 147640), specifically interacts with TLR3 (MIM 603029), and activates nuclear factor kappa-B (NFKB; see MIM 164011).[supplied by OMIM
Specifications
| TICAM1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TICAM1. | |
| In PBS (0.02% sodium azide) | |
| MGC35334/PRVTIRB/TICAM-1/TRIF | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| TICAM1 | |
| TICAM1 | |
| A synthetic peptide corresponding to C-terminus 14 amino acids of human TICAM1. | |
| RUO | |
| 148022 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only