missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TICAM2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TICAM2.
Brand: Abnova PAB13794.50ug
This item is not returnable.
View return policy
Description
TIRP is a Toll/interleukin-1 receptor (IL1R; MIM 147810) (TIR) domain-containing adaptor protein involved in Toll receptor signaling (see TLR4; MIM 603030).[supplied by OMIM
Sequence: GIGKSKINSCPLSLSWGKRHSVDTSPGYSpecifications
| TICAM2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TICAM2. | |
| In serum (0.02% sodium azide) | |
| MGC129876/MGC129877/TICAM-2/TIRAP3/TIRP/TRAM | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Recognizes human TRAM. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| TICAM2 | |
| TICAM2 | |
| A synthetic peptide corresponding to amino acids 2-29 of human TICAM2. | |
| RUO | |
| 353376 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction